Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343715.1 | NP_001330213.1 | 8 - 9 | 1144 | 116 | |
NM_147883.6 | NP_680188.1 | 8 - 9 | 1144 | 116 | ||
GenBank mRNA | BT014965.1 | - | 6 - 7 | 942 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |