Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334207.1 | NP_001320342.1 | 3 - 4 | 444 | 69 | |
NM_105222.3 | NP_176726.1 | 3 - 4 | 389 | 69 | ||
GenBank mRNA | AB027504.1 | - | 3 - 4 | 376 | 69 | |
AY065378.1 | - | 3 - 4 | 389 | 69 | ||
AY133813.1 | - | 3 - 4 | 307 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Inventoried Probe spans exons |