Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEXO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105233.5 | NP_176737.2 | 10 - 11 | 1472 | 115 | |
GenBank mRNA | AY128283.1 | - | 10 - 11 | 1046 | 115 | |
BT000831.1 | - | 10 - 11 | 1028 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |