Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YSL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105247.4 | NP_176750.1 | 1 - 2 | 629 | 110 | |
GenBank mRNA | AY515565.1 | - | 1 - 2 | 489 | 110 | |
BT002954.1 | - | 1 - 2 | 417 | 110 | ||
BT006077.1 | - | 1 - 2 | 378 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Arabidopsis thaliana Probe spans exons |