Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105256.4 | NP_176759.1 | 2 - 3 | 575 | 70 | |
GenBank mRNA | AF364953.1 | - | 2 - 3 | 529 | 70 | |
BT000353.1 | - | 2 - 3 | 160 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |