Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001124084.1 | NP_001117556.1 | 2 - 3 | 374 | 119 | |
GenBank mRNA | AF428372.1 | - | 2 - 3 | 315 | 119 | |
AY124873.1 | - | 2 - 3 | 283 | 119 | ||
U46665.1 | - | 2 - 3 | 299 | 119 | ||
U49937.1 | - | 2 - 3 | 283 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Arabidopsis thaliana Probe spans exons |