Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105345.5 | NP_176847.1 | 1 - 2 | 360 | 105 | |
GenBank mRNA | AB074116.1 | - | 1 - 2 | 147 | 105 | |
AY136355.1 | - | 1 - 2 | 134 | 105 | ||
BT000198.1 | - | 1 - 2 | 108 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |