Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334271.1 | NP_001321234.1 | 4 - 5 | 1024 | 89 | |
NM_105378.4 | NP_564889.1 | 5 - 6 | 952 | 89 | ||
GenBank mRNA | AK175554.1 | - | 5 - 6 | 578 | 89 | |
AY084717.1 | - | 5 - 6 | 708 | 89 | ||
BT024604.1 | - | 5 - 6 | 530 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |