Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SWEETIE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198408.1 | NP_001185337.1 | 31 - 32 | 3406 | 68 | |
NM_001334274.1 | NP_001319331.1 | 31 - 32 | 3478 | 68 | ||
NM_105384.8 | NP_176885.7 | 31 - 32 | 3514 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |