Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105392.5 | NP_176892.1 | 1 - 2 | 545 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |