Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334301.1 | NP_001319333.1 | 13 - 14 | 3324 | 84 | |
NM_001334302.1 | NP_001320390.1 | 14 - 15 | 3297 | 84 | ||
NM_001334303.1 | NP_001320389.1 | 14 - 15 | 3278 | 84 | ||
NM_001334304.1 | NP_001320393.1 | 14 - 15 | 3274 | 84 | ||
NM_001334305.1 | NP_001320391.1 | 14 - 15 | 3276 | 84 | ||
NM_001334306.1 | NP_001320392.1 | 13 - 14 | 3365 | 84 | ||
NM_105417.4 | NP_176917.2 | 14 - 15 | 3249 | 84 | ||
GenBank mRNA | AB114052.1 | - | 13 - 14 | 3117 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |