Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105422.4 | NP_176922.1 | 16 - 17 | 2409 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |