Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105469.2 | NP_564917.1 | 4 - 5 | 452 | 128 | |
GenBank mRNA | AY088274.1 | - | 4 - 5 | 452 | 128 | |
BT026412.1 | - | 4 - 5 | 413 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |