Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105470.3 | NP_176967.1 | 2 - 3 | 369 | 93 | |
GenBank mRNA | AJ000539.1 | - | 2 - 3 | 206 | 93 | |
AY140047.1 | - | 2 - 3 | 261 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |