Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105470.3 | NP_176967.1 | 9 - 10 | 822 | 121 | |
GenBank mRNA | AJ000539.1 | - | 9 - 10 | 659 | 121 | |
AY140047.1 | - | 9 - 10 | 714 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |