Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198420.1 | NP_001185349.1 | 10 - 11 | 1204 | 64 | |
NM_001334339.1 | NP_001320607.1 | 10 - 11 | 1179 | 64 | ||
NM_105471.4 | NP_176968.1 | 10 - 11 | 1204 | 64 | ||
GenBank mRNA | AF370221.1 | - | 10 - 11 | 1044 | 64 | |
AK226564.1 | - | 1021 | 64 | |||
AY099552.1 | - | 10 - 11 | 1036 | 64 | ||
AY113871.1 | - | 10 - 11 | 979 | 64 | ||
BT001237.1 | - | 10 - 11 | 979 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |