Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP70-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105476.5 | NP_176973.1 | 2 - 3 | 701 | 76 | |
GenBank mRNA | AM086438.1 | - | 2 - 3 | 326 | 76 | |
AY074303.1 | - | 2 - 3 | 599 | 76 | ||
AY096658.1 | - | 2 - 3 | 326 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |