Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334346.1 | NP_001322211.1 | 5 - 6 | 840 | 94 | |
NM_105480.3 | NP_564921.1 | 5 - 6 | 849 | 94 | ||
GenBank mRNA | AF216524.1 | - | 5 - 6 | 674 | 94 | |
AK229148.1 | - | 5 - 6 | 731 | 94 | ||
AY084533.1 | - | 5 - 6 | 745 | 94 | ||
BT001920.1 | - | 5 - 6 | 624 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |