Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334346.1 | NP_001322211.1 | 7 - 8 | 1215 | 75 | |
NM_105480.3 | NP_564921.1 | 7 - 8 | 1224 | 75 | ||
GenBank mRNA | AF216524.1 | - | 7 - 8 | 1049 | 75 | |
AK229148.1 | - | 7 - 8 | 1106 | 75 | ||
AY084533.1 | - | 7 - 8 | 1120 | 75 | ||
BT001920.1 | - | 7 - 8 | 999 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |