Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105508.4 | NP_177004.1 | 3 - 4 | 587 | 148 | |
GenBank mRNA | AF089810.1 | - | 3 - 4 | 495 | 148 | |
BT025556.1 | - | 3 - 4 | 334 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |