Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XYL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105527.5 | NP_177023.1 | 1 - 2 | 454 | 72 | |
GenBank mRNA | AY057482.1 | - | 1 - 2 | 315 | 72 | |
BT002675.1 | - | 1 - 2 | 217 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |