Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105548.5 | NP_177043.2 | 12 - 13 | 1421 | 106 | |
GenBank mRNA | AJ532903.1 | - | 12 - 13 | 1378 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |