Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105557.6 | NP_564946.1 | 2 - 3 | 669 | 97 | |
GenBank mRNA | BT030352.1 | - | 2 - 3 | 480 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |