Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105585.3 | NP_177078.1 | 5 - 6 | 658 | 77 | |
GenBank mRNA | AF132606.1 | - | 5 - 6 | 481 | 77 | |
AK229609.1 | - | 5 - 6 | 536 | 77 | ||
BT008618.1 | - | 5 - 6 | 536 | 77 | ||
DQ446412.1 | - | 5 - 6 | 457 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |