Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105604.2 | NP_177096.1 | 5 - 6 | 854 | 77 | |
GenBank mRNA | AK117860.1 | - | 5 - 6 | 728 | 77 | |
BT005306.1 | - | 5 - 6 | 712 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |