Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT1G69460 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105613.5 | NP_177105.2 | 3 - 4 | 601 | 149 | |
GenBank mRNA | AY072343.1 | - | 3 - 4 | 539 | 149 | |
AY114598.1 | - | 3 - 4 | 505 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |