Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SBH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105632.5 | NP_177122.2 | 1 - 2 | 487 | 60 | |
GenBank mRNA | AY070765.1 | - | 1 - 2 | 334 | 60 | |
AY143918.1 | - | 1 - 2 | 227 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |