Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEMB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084331.3 | NP_001077800.1 | 10 - 11 | 1188 | 100 | |
NM_001334421.1 | NP_001320950.1 | 9 - 10 | 1302 | 100 | ||
NM_105642.4 | NP_177132.1 | 9 - 10 | 1214 | 100 | ||
GenBank mRNA | AF327428.1 | - | 9 - 10 | 922 | 100 | |
AF361803.1 | - | 9 - 10 | 958 | 100 | ||
AY059154.1 | - | 8 - 9 | 893 | 100 | ||
AY081254.1 | - | 9 - 10 | 964 | 100 | ||
AY113941.1 | - | 8 - 9 | 893 | 100 | ||
AY120705.1 | - | 9 - 10 | 922 | 100 | ||
AY128711.1 | - | 8 - 9 | 893 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |