Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105645.4 | NP_177135.1 | 17 - 18 | 2304 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |