Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105651.5 | NP_564977.1 | 2 - 3 | 581 | 111 | |
GenBank mRNA | AY050398.1 | - | 2 - 3 | 457 | 111 | |
BT000643.1 | - | 2 - 3 | 411 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |