Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KUP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105698.5 | NP_177187.2 | 5 - 6 | 1676 | 90 | |
GenBank mRNA | AY062542.1 | - | 5 - 6 | 1371 | 90 | |
AY093328.1 | - | 5 - 6 | 1279 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |