Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105734.3 | NP_564994.1 | 4 - 5 | 582 | 80 | |
GenBank mRNA | BT004011.1 | - | 4 - 5 | 567 | 80 | |
BT004943.1 | - | 4 - 5 | 422 | 80 |
Gene Symbol: AT1G70650 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198435.1 | NP_001185364.1 | 9 - 10 | 1813 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |