Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GH9B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105739.4 | NP_177228.1 | 2 - 3 | 996 | 76 | |
GenBank mRNA | AY048283.1 | - | 2 - 3 | 659 | 76 | |
AY074552.1 | - | 2 - 3 | 621 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |