Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GH9B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105739.4 | NP_177228.1 | 4 - 5 | 1245 | 92 | |
GenBank mRNA | AY048283.1 | - | 4 - 5 | 908 | 92 | |
AY074552.1 | - | 4 - 5 | 870 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Arabidopsis thaliana Probe spans exons |