Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GH9B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105739.4 | NP_177228.1 | 5 - 6 | 1358 | 107 | |
GenBank mRNA | AY048283.1 | - | 5 - 6 | 1021 | 107 | |
AY074552.1 | - | 5 - 6 | 983 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |