Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105762.3 | NP_177250.1 | 1 - 2 | 1461 | 95 | |
GenBank mRNA | AF087818.1 | - | 1 - 2 | 1336 | 95 | |
AK226890.1 | - | 1 - 2 | 1315 | 95 | ||
AY136327.1 | - | 1 - 2 | 1347 | 95 | ||
BT002085.1 | - | 1 - 2 | 1231 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |