Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105762.3 | NP_177250.1 | 4 - 5 | 2014 | 67 | |
GenBank mRNA | AF087818.1 | - | 4 - 5 | 1889 | 67 | |
AK226890.1 | - | 4 - 5 | 1868 | 67 | ||
AY136327.1 | - | 4 - 5 | 1900 | 67 | ||
BT002085.1 | - | 4 - 5 | 1784 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |