Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105807.2 | NP_177294.1 | 4 - 5 | 1190 | 93 | |
GenBank mRNA | BT026459.1 | - | 4 - 5 | 963 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |