Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105825.3 | NP_565022.1 | 5 - 6 | 685 | 144 | |
GenBank mRNA | AK228190.1 | - | 5 - 6 | 685 | 144 | |
BT006157.1 | - | 5 - 6 | 683 | 144 | ||
BT008332.1 | - | 5 - 6 | 497 | 144 | ||
BT008524.1 | - | 5 - 6 | 497 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |