Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198445.1 | NP_001185374.1 | 14 - 15 | 1579 | 100 | |
NM_105840.4 | NP_565026.1 | 14 - 15 | 1432 | 100 | ||
GenBank mRNA | AY046014.1 | - | 14 - 15 | 1353 | 100 | |
AY133819.1 | - | 13 - 14 | 1084 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |