Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198445.1 | NP_001185374.1 | 17 - 18 | 1917 | 62 | |
NM_105840.4 | NP_565026.1 | 17 - 18 | 1836 | 62 | ||
GenBank mRNA | AY046014.1 | - | 17 - 18 | 1757 | 62 | |
AY133819.1 | - | 16 - 17 | 1488 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |