Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198445.1 | NP_001185374.1 | 24 - 25 | 2491 | 109 | |
NM_105840.4 | NP_565026.1 | 24 - 25 | 2410 | 109 | ||
GenBank mRNA | AY046014.1 | - | 24 - 25 | 2331 | 109 | |
AY133819.1 | - | 23 - 24 | 2062 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |