Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036189.1 | NP_001031266.1 | 2 - 3 | 448 | 125 | |
NM_105844.4 | NP_177331.1 | 2 - 3 | 482 | 125 | ||
NM_202398.1 | NP_974127.1 | 2 - 3 | 430 | 125 | ||
GenBank mRNA | AY070403.1 | - | 2 - 3 | 430 | 125 | |
AY117240.1 | - | 2 - 3 | 293 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |