Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105846.4 | NP_177333.1 | 1 - 2 | 1559 | 90 | |
GenBank mRNA | AY049275.1 | - | 1 - 2 | 1409 | 90 | |
BT020416.1 | - | 1 - 2 | 1255 | 90 | ||
X75365.1 | - | 1314 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |