Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105857.4 | NP_177344.2 | 1 - 2 | 598 | 71 | |
GenBank mRNA | AJ404862.1 | - | 1 - 2 | 463 | 71 | |
BT026431.1 | - | 1 - 2 | 463 | 71 | ||
KJ138942.1 | - | 1 - 2 | 463 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |