Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036190.2 | NP_001031267.1 | 3 - 4 | 701 | 76 | |
NM_105896.4 | NP_177381.1 | 3 - 4 | 701 | 76 | ||
GenBank mRNA | AK316989.1 | - | 3 - 4 | 677 | 76 | |
AY054211.1 | - | 3 - 4 | 677 | 76 | ||
AY058885.1 | - | 3 - 4 | 697 | 76 | ||
AY065096.1 | - | 3 - 4 | 694 | 76 | ||
AY079041.1 | - | 3 - 4 | 612 | 76 | ||
AY087976.1 | - | 3 - 4 | 694 | 76 | ||
AY114561.1 | - | 3 - 4 | 612 | 76 | ||
AY136324.1 | - | 3 - 4 | 674 | 76 | ||
BT000421.1 | - | 3 - 4 | 612 | 76 | ||
U01955.1 | - | 3 - 4 | 666 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |