Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105930.3 | NP_177415.1 | 13 - 14 | 1473 | 70 | |
GenBank mRNA | AY059850.1 | - | 13 - 14 | 1399 | 70 | |
AY943843.1 | - | 13 - 14 | 1117 | 70 | ||
BT010328.1 | - | 13 - 14 | 1117 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |