Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NF-YA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036195.5 | NP_001031272.1 | 3 - 4 | 816 | 113 | |
NM_001334553.1 | NP_001319368.1 | 3 - 4 | 693 | 113 | ||
NM_105941.8 | NP_565049.1 | 3 - 4 | 1069 | 113 | ||
GenBank mRNA | AY140028.1 | - | 3 - 4 | 586 | 113 | |
BT008825.1 | - | 2 - 3 | 392 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |