Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198461.1 | NP_001185390.1 | 10 - 11 | 2382 | 138 | |
NM_001334591.1 | NP_001319374.1 | 11 - 12 | 2693 | 138 | ||
NM_105997.2 | NP_177480.1 | 10 - 11 | 2629 | 138 | ||
GenBank mRNA | KJ138940.1 | - | 10 - 11 | 2266 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |