Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198461.1 | NP_001185390.1 | 8 - 9 | 1597 | 113 | |
NM_001334591.1 | NP_001319374.1 | 9 - 10 | 1908 | 113 | ||
NM_105997.2 | NP_177480.1 | 8 - 9 | 1844 | 113 | ||
GenBank mRNA | KJ138940.1 | - | 8 - 9 | 1481 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |